Läs mer
1-[(1RS)-1-(4-Methoxyphenyl)-2-(methylamino)ethyl]cyclohexanol Hydrochloride, Mikromol™
Beskrivning
1-[(1RS)-1-(4-Methoxyphenyl)-2-(methylamino)ethyl]cyclohexanol Hydrochloride
Specifikationer
Specifikationer
| Kemiskt namn eller material | N-Desmethylvenlafaxine Hydrochloride |
| CAS | 93413-90-2 |
| Analyt- eller komponentnamn | N-Desmethylvenlafaxine Hydrochloride |
| Frakt skick | Room Temperature |
| Molekylformel | C16 H25 N O2 . Cl H |
| Certifikat och efterlevnad | ISO 17025 |
| Synonym | Venlafaxine Hydrochloride Imp. D (EP), Venlafaxine Imp. D (EP), Venlafaxine USP Related Compound A, Venlafaxine USP RC A, 1-[(1RS)-1-(4-Methoxyphenyl)-2-(methylamino)ethyl]cyclohexanol Hydrochloride, Venlafaxine Hydrochloride Imp. D (EP) as Hydrochloride, N-Desmethylvenlafaxine Hydrochloride, Venlafaxine Related Compound A as Hydrochloride, Venlafaxine Hydrochloride Impurity D as Hydrochloride, Venlafaxine Impurity D as Hydrochloride |
| InChI formel | InChI=1S/C16H25NO2.ClH/c1-17-12-15(16(18)10-4-3-5-11-16)13-6-8-14(19-2)9-7-13;/h6-9,15,17-18H,3-5,10-12H2,1-2H3;1H |
| LEDER | Cl.CNCC(c1ccc(OC)cc1)C2(O)CCCCC2 |
| IUPAC-namn | 1-[1-(4-methoxyphenyl)-2-(methylamino)ethyl]cyclohexan-1-ol;hydrochloride |
| Visa mer |