Läs mer
1-[2-(2,4-Dichlorophenyl)-2-[(3,4-dichlorophenyl)methoxy]ethyl]-1H-imidazole, TRC
Produktkod. 30011881
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30011881 100 mg
30005515 10 mg
30010462 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30011881 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD432038100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 47447-55-2
Molekylformel C18H14Cl4N2O
Molekylvikt (g/mol) 416.13
Synonym Miconazole EP Impurity F;
LEDER Clc1ccc(C(Cn2ccnc2)OCc3ccc(Cl)c(Cl)c3)c(Cl)c1
Specifikationer
Kemiskt namn eller material 1-[2-(2,4-Dichlorophenyl)-2-[(3,4-dichlorophenyl)methoxy]ethyl]-1H-imidazole
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C18H14Cl4N2O/c19-13-2-3-14(16(21)8-13)18(9-24-6-5-23-11-24)25-10-12-1-4-15(20)17(22)7-12/h1-8,11,18H,9-10H2
Formel vikt 413.986
Procent renhet >95%