missing translation for 'onlineSavingsMsg'
Learn More

1,2,2,6,6-Pentamethylpiperidine, 97%, Thermo Scientific Chemicals

Catalog Number p-7030182
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
79-55-0
C10H21N
155.285
MFCD00006493
XULIXFLCVXWHRF-UHFFFAOYSA-N
pempidine, pyrilene, perolysen, tenormal, piperidine, 1,2,2,6,6-pentamethyl, pempidina dcit, pempidine inn:ban, pempidinum inn-latin, unii-n5i18ji9d6, 1,2,2,6,6-pentamethyl-piperidine
6603
1,2,2,6,6-pentamethylpiperidine
CC1(CCCC(N1C)(C)C)C
This item is not returnable. View return policy
79-55-0
C10H21N
155.285
MFCD00006493
XULIXFLCVXWHRF-UHFFFAOYSA-N
pempidine, pyrilene, perolysen, tenormal, piperidine, 1,2,2,6,6-pentamethyl, pempidina dcit, pempidine inn:ban, pempidinum inn-latin, unii-n5i18ji9d6, 1,2,2,6,6-pentamethyl-piperidine
6603
1,2,2,6,6-pentamethylpiperidine
CC1(CCCC(N1C)(C)C)C

Pharmacological evaluation of the antagonism of nicotine's central effects by mecamylamine and pempidine where pempidine was found to increase the ED50 of nicotine (0.73 mg/kg) for depression of spontaneous activity in a dose-related manner. At a dose of 3 mg/kg, pempidine increased nicotine's ED50 4.7-fold.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pharmacological evaluation of the antagonism of nicotine′s central effects by mecamylamine and pempidine where pempidine was found to increase the ED50 of nicotine (0.73 mg/kg) for depression of spontaneous activity in a dose-related manner. At a dose of 3 mg/kg, pempidine increased nicotine′s ED50 4.7-fold.

Solubility
Soluble in toluene, dimethylformamide.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
TRUSTED_SUSTAINABILITY
Density 0.858
Boiling Point 187°C to 188°C
Flash Point 50°C (122°F)
Odor Amine-like
Quantity 1 g
UN Number UN1992
Beilstein 103806
Sensitivity Air Sensitive
Merck Index 14,7080
Solubility Information Soluble in toluene,dimethylformamide.
Formula Weight 155.28
Percent Purity 97%
Chemical Name or Material 1,2,2,6,6-Pentamethylpiperidine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'