Läs mer
1-[[2-[(2-Aminophenyl)amino]-5-methyl-3-thienyl]carbonyl]-4-methyl-piperazine, TRC
Produktkod. 30134948
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134948 500 mg
30003277 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30134948 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA622450500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 138564-61-1
Molekylformel C17H22N4OS
Molekylvikt (g/mol) 330.45
Synonym [2-[(2-Aminophenyl)amino]-5-methyl-3-thienyl](4-methyl-1-piperazinyl)methanone, 1-[[2-[(2-Aminophenyl)amino]-5-methyl-3-thienyl]carbonyl]-4-methylpiperazine (9CI)
IUPAC-namn [2-(2-aminoanilino)-5-methylthiophen-3-yl]-(4-methylpiperazin-1-yl)methanone
LEDER CN1CCN(CC1)C(=O)c2cc(C)sc2Nc3ccccc3N
Specifikationer
Kemiskt namn eller material 1-[[2-[(2-Aminophenyl)amino]-5-methyl-3-thienyl]carbonyl]-4-methyl-piperazine
Analyt- eller komponentnamn 1-[[2-[(2-Aminophenyl)amino]-5-methyl-3-thienyl]carbonyl]-4-methyl-piperazine
Frakt skick Room Temperature
InChI formel InChI=1S/C17H22N4OS/c1-12-11-13(17(22)21-9-7-20(2)8-10-21)16(23-12)19-15-6-4-3-5-14(15)18/h3-6,11,19H,7-10,18H2,1-2H3
Lösningstyp Neat
Industry Type Pharmaceutical
Kvantitet 500 mg
Formel vikt 330.1514