Läs mer
1,2,3,4,5,6-Benzenehexamine Trihydrochloride, TRC
Produktkod. 30006852
Change view
Kvantitet:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30006852 50 mg
30005788 250 mg
30010077 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30006852 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB18990550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 126521-82-2
Molekylformel C6H12N6 • 3HCl
Molekylvikt (g/mol) 168.2
Synonym Hexaaminobenzene Trihydrochloride, Benzenehexamine Trihydrochloride
IUPAC-namn benzene-1,2,3,4,5,6-hexaamine
LEDER NC1=C(N)C(N)=C(N)C(N)=C1N
Specifikationer
Kemiskt namn eller material 1,2,3,4,5,6-Benzenehexamine Trihydrochloride
Analyt- eller komponentnamn 1,2,3,4,5,6-Benzenehexamine Trihydrochloride
Anteckningar om renhetsgrad HPLC
Frakt skick Room Temperature
InChI formel InChI=1S/C6H12N6.3ClH/c7-1-2(8)4(10)6(12)5(11)3(1)9;;;/h7-12H2;3*1H
Lösningstyp Neat
Industry Type Pharmaceutical
Kvantitet 50 mg
Formel vikt 168.1123
Procent renhet >95%