Läs mer
1,2,3,4,5-Pentanepentacarboxylic Acid Sodium Salt Trihydrate, TRC
Produktkod. 30427596
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30427596 10 mg
30414481 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30427596 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP27350010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 55525-23-0
Molekylformel C10H13Na5O13
Molekylvikt (g/mol) 456.15
Synonym Pentasodium Pentane-1,2,3,4,5-pentacarboxylic Acid Trihydrate, sodium pentane-1,2,3,4,5-pentacarboxylate trihydrate
IUPAC-namn sodium pentane-1,2,3,4,5-pentacarboxylate trihydrate
LEDER O=C(CC(C(O[Na])=O)C(C(O[Na])=O)C(C(O[Na])=O)CC(O[Na])=O)O[Na].O.O.O
Specifikationer
Kemiskt namn eller material 1,2,3,4,5-Pentanepentacarboxylic Acid Sodium Salt Trihydrate
Kvantitet 10 mg
InChI formel InChI=1S/C10H12O10.5Na.3H2O/c11-5(12)1-3(8(15)16)7(10(19)20)4(9(17)18)2-6(13)14;;;;;;;;/h3-4,7H,1-2H2,(H,11,12)(H,13,14)(H,15,16)(H,17,18)(H,19,20);;;;;;3*1H2/q;5*+1;;;/p-5
Formel vikt 455.98