Läs mer
1,2,3,4,6-Pentachloronaphthalene, TRC
Produktkod. 30428674
Change view
Kvantitet:
2.5 mg
10 mg
25 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30428674 2.5 mg 2.5mg
30423566 25 mg -
30426017 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30428674 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP2375302.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 67922-26-3
Molekylformel C10 H3 Cl5
Molekylvikt (g/mol) 300.4
Synonym Naphthalene, 1,2,3,4,6-pentachloro-, 1,2,3,4,6-Pentachloronaphthalene, PCN 50, 1,2,3,4,6-PentaCN, 1,2,3,4,6-PCN, 12346-Pentachloronaphthalene, 12346-PentaCN, 12346-PCN, PCN-50, PCN No. 50
IUPAC-namn 1,2,3,4,6-pentachloronaphthalene
LEDER Clc1ccc2c(Cl)c(Cl)c(Cl)c(Cl)c2c1
Specifikationer
Kemiskt namn eller material 1,2,3,4,6-Pentachloronaphthalene
Anteckningar om renhetsgrad HPLC
Kvantitet 2.5 mg
InChI formel InChI=1S/C10H3Cl5/c11-4-1-2-5-6(3-4)8(13)10(15)9(14)7(5)12/h1-3H
Formel vikt 297.8677
Procent renhet >95%