Läs mer
1,2,3,4,7,8-Hexachlorodibenzofuran (>90%), TRC
Produktkod. 30137824
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137824 50 mg
30005944 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30137824 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH26509550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 70648-26-9
Molekylformel C12 H2 Cl6 O
Molekylvikt (g/mol) 374.86
Synonym Dibenzofuran, 1,2,3,4,7,8-hexachloro-, 1,2,3,4,7,8-Hexachlorodibenzofuran, 1,2,3,4,7,8-HCDF, 1,2,3,4,7,8-HxCDF, 123478-HxCDF, F 118, PCDF 118, 1,2,3,4,7,8-HexaCDF, 123478-HexaCDF, 123478-Hexachlorodibenzofuran
IUPAC-namn 1,2,3,4,7,8-hexachlorodibenzofuran
LEDER Clc1cc2oc3c(Cl)c(Cl)c(Cl)c(Cl)c3c2cc1Cl
Specifikationer
Kemiskt namn eller material 1,2,3,4,7,8-Hexachlorodibenzofuran
Kvantitet 50 mg
InChI formel InChI=1S/C12H2Cl6O/c13-4-1-3-6(2-5(4)14)19-12-7(3)8(15)9(16)10(17)11(12)18/h1-2H
Formel vikt 371.8237