Läs mer
1,2,3,4-Tetra-O-acetyl-L-fucopyranose, 90%, TRC
Produktkod. 30424492
Change view
Kvantitet:
1 g
2 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30424492 2 g
30419592 1 g
30418449 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30424492 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT2792552G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 24332-95-4
Molekylformel C14 H20 O9
Molekylvikt (g/mol) 332.3
Synonym 1,2,3,4-Tetra-O-Acetyl-L-Fucopyranose
IUPAC-namn [(2S,3R,4R,5S)-4,5,6-triacetyloxy-2-methyloxan-3-yl] acetate
LEDER C[C@@H]1OC(OC(=O)C)[C@@H](OC(=O)C)[C@H](OC(=O)C)[C@@H]1OC(=O)C
Specifikationer
Kemiskt namn eller material 1,2,3,4-Tetra-O-acetyl-L-fucopyranose
Kvantitet 2 g
InChI formel InChI=1S/C14H20O9/c1-6-11(20-7(2)15)12(21-8(3)16)13(22-9(4)17)14(19-6)23-10(5)18/h6,11-14H,1-5H3/t6-,11+,12+,13-,14?/m0/s1
Formel vikt 332.1107