Läs mer
1,2,3,4-Tetrachloronaphthalene, TRC
Produktkod. 30453547
Change view
Kvantitet:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30453547 50 mg
30449953 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30453547 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT31063550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1,2,3,4-Tetrachloronaphthalene
CAS 20020-02-4
Molekylformel C10 H4 Cl4
Kvantitet 50 mg
Synonym Naphthalene','1,2,3,4-tetrachloro-, 1,2,3,4-Tetrachloronaphthalene, NSC 524443, PCN 27, TCN, 1,2,3,4-TetraCN, 1,2,3,4-TeCN, 1234-Tetrachloronaphthalene, 1234-TetraCN, 1234-TeCN, PCN-27
InChI formel InChI=1S/C10H4Cl4/c11-7-5-3-1-2-4-6(5)8(12)10(14)9(7)13/h1-4H
LEDER Clc1c(Cl)c(Cl)c2ccccc2c1Cl
IUPAC-namn 1,2,3,4-tetrachloronaphthalene
Molekylvikt (g/mol) 265.95
Formel vikt 263.9067