Läs mer
1,2,3,4-Tetrahydro-2-[2-(4-hydroxyphenyl)-1-methylethyl]-4,6,8-isoquinolinetriol, TRC
Produktkod. 30136772
Change view
Kvantitet:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136772 5 mg
30136457 10 mg
17967444 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136772 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT2943055MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 161040-25-1
Molekylformel C18 H21 N O4
Molekylvikt (g/mol) 315.36
Synonym 1,2,3,4-Tetrahydro-2-[2-(4-hydroxyphenyl)-1-methylethyl]-4,6,8-isoquinolinetriol
IUPAC-namn 2-[1-(4-hydroxyphenyl)propan-2-yl]-3,4-dihydro-1H-isoquinoline-4,6,8-triol
LEDER CC(Cc1ccc(O)cc1)N2CC(O)c3cc(O)cc(O)c3C2
Specifikationer
Kemiskt namn eller material 1,2,3,4-Tetrahydro-2-[2-(4-hydroxyphenyl)-1-methylethyl]-4,6,8-isoquinolinetriol
Anteckningar om renhetsgrad HPLC
Kvantitet 5 mg
InChI formel InChI=1S/C18H21NO4/c1-11(6-12-2-4-13(20)5-3-12)19-9-16-15(18(23)10-19)7-14(21)8-17(16)22/h2-5,7-8,11,18,20-23H,6,9-10H2,1H3
Formel vikt 315.1471
Procent renhet >95%