Läs mer
1,2,3,4-Tetrahydro-4,6-dihydroxy-2-methyl-isoquinoline, TRC
Produktkod. 30012911
Change view
Kvantitet:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30012911 50 mg
30005586 250 mg
30012306 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30012911 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT29357550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 23824-24-0
Molekylformel C10 H13 N O2
Molekylvikt (g/mol) 179.22
Synonym 4,6-Isoquinolinediol, 1,2,3,4-tetrahydro-2-methyl-, 1,2,3,4-Tetrahydro-2-methyl-4,6-isoquinolinediol, 1,2,3,4-Tetrahydro-4,6-dihydroxy-2-methylisoquinoline, 2-Methyl-1,2,3,4-tetrahydroisoquinoline-4,6-diol, 4,6-Dihydroxy-2-methyl-1,2,3,4-tetrahydroisoquinoline
IUPAC-namn 2-methyl-3,4-dihydro-1H-isoquinoline-4,6-diol
LEDER CN1CC(O)c2cc(O)ccc2C1
Specifikationer
Kemiskt namn eller material 1,2,3,4-Tetrahydro-4,6-dihydroxy-2-methyl-isoquinoline
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C10H13NO2/c1-11-5-7-2-3-8(12)4-9(7)10(13)6-11/h2-4,10,12-13H,5-6H2,1H3
Formel vikt 179.0946
Procent renhet >95%