Läs mer
1,2,3,6-Tetrahydropyridine Hydrochloride ~90%, TRC
Produktkod. 30452820
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30452820 1 mg
1 options
Produktkod. 30452820 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT7947111MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1,2,3,6-Tetrahydropyridine Hydrochloride
CAS 18513-79-6
Molekylformel C5 H9 N . H Cl
Kvantitet 1 mg
Synonym Pyridine','1,2,3,6-tetrahydro-','hydrochloride (1:1), Pyridine','1,2,3,6-tetrahydro-','hydrochloride (7CI,8CI,9CI), 1,2,3,6-Tetrahydropyridine hydrochloride, 1,2,5,6-Tetrahydropyridine hydrochloride, 3,6-Dihydro-2H-pyridine hydrochloride, Δ3-Piperidine hydrochloride
InChI formel InChI=1S/C5H9N.ClH/c1-2-4-6-5-3-1;/h1-2,6H,3-5H2;1H
LEDER Cl.C1CC=CCN1
IUPAC-namn 1,2,3,6-tetrahydropyridine;hydrochloride
Molekylvikt (g/mol) 119.593
Formel vikt 119.05