Läs mer
1,2,3,7,8,9-Hexachlorodibenzodioxin, TRC
Produktkod. 30179785
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30179785 1 mg
30180185 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30179785 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH2907901MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 19408-74-3
Synonym Dibenzo[b,e][1,4]dioxin, 1,2,3,7,8,9-hexachloro-∣Dibenzo-p-dioxin, 1,2,3,7,8,9-hexachloro- (8CI)∣1,2,3,7,8,9-Hexachlorodibenzo[b,e][1,4]dioxin∣1,2,3,7,8,9-Hexachlorodibenzo-p-dioxin∣1,2,3,7,8,9-Hexachlorodibenzo[1,4]dioxin∣1,2,3,7,8,9-Hexachlorodibenzodioxin∣1,2,3,7,8,9-HxCDD∣123789-HxCDD∣D 70∣PCDD 70
Specifikationer
Formatera Neat
Molekylformel C12H2Cl6O2
Molekylvikt (g/mol) 390.86, 390.85
InChI-nyckel LGIRBUBHIWTVCK-UHFFFAOYSA-N
LEDER ClC1=CC2=C(OC3=C(Cl)C(Cl)=C(Cl)C=C3O2)C(Cl)=C1Cl
IUPAC-namn 1,2,3,7,8,9-hexachlorooxanthrene