Läs mer
1,2,3,7,8,9-Hexachlorodibenzofuran, TRC
Produktkod. 30408192
Change view
Kvantitet:
0.5 mg
2.5 mg
Förpackningsstorlek:
0.5mg
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30408192 0.5 mg 0.5mg
30392445 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30408192 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH2650900.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 72918-21-9
Molekylformel C12 H2 Cl6 O
Molekylvikt (g/mol) 374.86
Synonym Dibenzofuran, 1,2,3,7,8,9-hexachloro-, 1,2,3,7,8,9-Hexachlorodibenzofuran, 1,2,3,7,8,9-HxCDF, F 124, PCDF 124, 1,2,3,7,8,9-HexaCDF, 123789-HexaCDF, 123789-HxCDF, 123789-Hexachlorodibenzofuran
IUPAC-namn 1,2,3,7,8,9-hexachlorodibenzofuran
LEDER Clc1cc2oc3cc(Cl)c(Cl)c(Cl)c3c2c(Cl)c1Cl
Specifikationer
Kemiskt namn eller material 1,2,3,7,8,9-Hexachlorodibenzofuran
Kvantitet 0.5 mg
InChI formel InChI=1S/C12H2Cl6O/c13-3-1-5-7(11(17)9(3)15)8-6(19-5)2-4(14)10(16)12(8)18/h1-2H
Formel vikt 371.8237