Läs mer
1,2,3,7,8-Pentachlorodibenzofuran, TRC
Produktkod. 30138600
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138600 50 mg
18040190 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30138600 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP23797050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 57117-41-6
Molekylformel C12 H3 Cl5 O
Molekylvikt (g/mol) 340.42
Synonym Dibenzofuran, 1,2,3,7,8-pentachloro-, 1,2,3,7,8-Pentachlorodibenzofuran, 1,2,3,7,8-PeCDF, 12378-PCDF, F 94, PCDF 94, 1,2,3,7,8-PentaCDF, 1,2,3,7,8-PCDF, 12378-PentaCDF, 12378-Pentachlorodibenzofuran
IUPAC-namn 1,2,3,7,8-pentachlorodibenzofuran
LEDER Clc1cc2oc3cc(Cl)c(Cl)c(Cl)c3c2cc1Cl
Specifikationer
Kemiskt namn eller material 1,2,3,7,8-Pentachlorodibenzofuran
Kvantitet 50 mg
InChI formel InChI=1S/C12H3Cl5O/c13-5-1-4-8(2-6(5)14)18-9-3-7(15)11(16)12(17)10(4)9/h1-3H
Formel vikt 337.8627