Läs mer
1-(2,3-Di-O-acetyl-5-deoxy-beta-D-ribofuranosyl)-4-amino-5-fluoropyrimidin-2(1H)-one, Mikromol™
Produktkod. 30052138
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30052138 100 mg
30051594 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30052138 Leverantör Mikromol Leverantörsnummer MM0992.07
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1-(2,3-Di-O-acetyl-5-deoxy-beta-D-ribofuranosyl)-4-amino-5-fluoropyrimidin-2(1H)-one
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2',3'-Di-O-acetyl-5'-deoxy-5-fluorocytidine
CAS 161599-46-8
Analyt- eller komponentnamn 2',3'-Di-O-acetyl-5'-deoxy-5-fluorocytidine
Frakt skick Room Temperature
Molekylformel C13 H16 F N3 O6
Certifikat och efterlevnad ISO 17025
Synonym 5'-Deoxy-5-fluorocytidine 2',3'-Diacetate, 2',3'-Di-O-acetyl-5'-deoxy-5-fluorocytidine, 5'-Deoxy-2',3'-di-O-acetyl-5-fluorocytidine, Capecitabine Imp. C (EP), Capecitabine Impurity C
InChI formel InChI=1S/C13H16FN3O6/c1-5-9(22-6(2)18)10(23-7(3)19)12(21-5)17-4-8(14)11(15)16-13(17)20/h4-5,9-10,12H,1-3H3,(H2,15,16,20)/t5-,9-,10-,12-/m1/s1
LEDER C[C@H]1O[C@H]([C@H](OC(=O)C)[C@@H]1OC(=O)C)N2C=C(F)C(=NC2=O)N
IUPAC-namn [(2R,3R,4R,5R)-4-acetyloxy-5-(4-amino-5-fluoro-2-oxopyrimidin-1-yl)-2-methyloxolan-3-yl] acetate
Molekylvikt (g/mol) 329.28
Kvantitet 100 mg
Formel vikt 329.1023 g/mol