missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(2,3-Dichlorophenyl)-3-[4-(trifluoromethoxy)phenyl]urea, 97%

Brand:  Alfa Aesar™ H64883.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15489226

  • 2950.30 SEK / 1g


Estimated Shipment date
from Supplier 06-03-2018
Add to basket

Specifications

Chemical Identifiers

1-(2,3-Dichlorophenyl)-3-[4-(trifluoromethoxy)phenyl]urea
365.13
MFCD04281090

Specifications

1-(2,3-Dichlorophenyl)-3-[4-(trifluoromethoxy)phenyl]urea
Solid
365.13
1g
97%
MFCD04281090
Videos
SDS