Läs mer
1-(2,3-Dimethylphenyl)ethanone, TRC
Produktkod. 30179905
Change view
Kvantitet:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30179905 250 mg
30180420 1 g
30179823 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30179905 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD481073250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2142-71-4
Molekylformel C10H12O
Molekylvikt (g/mol) 148.2
Synonym 1-(2,3-Dimethylphenyl)ethan-1-one, 1-(2,3-Dimethylphenyl)ethanone;
LEDER CC(=O)c1cccc(C)c1C
Specifikationer
Kemiskt namn eller material 1-(2,3-Dimethylphenyl)ethanone
Kvantitet 250 mg
InChI formel InChI=1S/C10H12O/c1-7-5-4-6-10(8(7)2)9(3)11/h4-6H,1-3H3
Formel vikt 148.0888