missing translation for 'onlineSavingsMsg'
Learn More

1,2,3-Thiadiazole-4-carboxylic acid, 97%, Thermo Scientific™

Catalog Number p-2694589
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
4100-13-4
C3H2N2O2S
130.121
MFCD00052099
HJZYBDPHAHGHAZ-UHFFFAOYSA-N
1,2,3-thiadiazole-4-carboxylic acid, 1,2,3 thiadiazole-4-carboxylic acid, thiadiazolecarboxylicacid, 4-carboxy-1,2,3-thiadiazole, 1,2,3-thiadiazole4-carboxylic acid, 1 pound not2 pound not3 thiadiazole-4-carboxylicacid
351418
thiadiazole-4-carboxylic acid
C1=C(N=NS1)C(=O)O
This item is not returnable. View return policy
4100-13-4
C3H2N2O2S
130.121
MFCD00052099
HJZYBDPHAHGHAZ-UHFFFAOYSA-N
1,2,3-thiadiazole-4-carboxylic acid, 1,2,3 thiadiazole-4-carboxylic acid, thiadiazolecarboxylicacid, 4-carboxy-1,2,3-thiadiazole, 1,2,3-thiadiazole4-carboxylic acid, 1 pound not2 pound not3 thiadiazole-4-carboxylicacid
351418
thiadiazole-4-carboxylic acid
C1=C(N=NS1)C(=O)O
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 130.13
Percent Purity 97%
Chemical Name or Material 1, 2, 3-Thiadiazole-4-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'