Läs mer
1,2,3-Tri-O-benzoyl-4-deoxy-Alpha-L-fucopyranose, TRC
Produktkod. 30456253
Change view
Kvantitet:
10 mg
Product Code. Kvantitet
30456253 10 mg
1 missing translation for 'options'
Product Code. 30456253 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCT76765010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Kemiskt namn eller material 1,2,3-Tri-O-benzoyl-4-deoxy-a-L-fucopyranose
CAS 132867-80-2
Molekylformel C27 H24 O7
Kvantitet 10 mg
Synonym 1,2,3-Tri-O-Benzoyl-4-Deoxy-α-L-Fucopyranose
InChI formel InChI=1S/C27H24O7/c1-18-17-22(32-24(28)19-11-5-2-6-12-19)23(33-25(29)20-13-7-3-8-14-20)27(31-18)34-26(30)21-15-9-4-10-16-21/h2-16,18,22-23,27H,17H2,1H3/t18-,22+,23-,27-/m0/s1
LEDER C[C@H]1C[C@@H](OC(=O)c2ccccc2)[C@H](OC(=O)c3ccccc3)[C@H](OC(=O)c4ccccc4)O1
IUPAC-namn [(2S,3S,4R,6S)-2,3-dibenzoyloxy-6-methyloxan-4-yl] benzoate
Molekylvikt (g/mol) 460.48
Formel vikt 460.1522