Läs mer
1,2,3-Trichloro-5-methylbenzene, TRC
Produktkod. 17960975
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17960975 5 mg
17924175 25 mg
17913855 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17960975 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT7743685MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 21472-86-6
Molekylformel C7 H5 Cl3
Molekylvikt (g/mol) 195.474
Synonym 1,2,3-Trichloro-5-methylbenzene (ACI), Toluene, 3,4,5-trichloro- (8CI), 3,4,5-Trichlorotoluene, 3,4,5-Trichlorotoluene
IUPAC-namn 1,2,3-trichloro-5-methylbenzene
LEDER Cc1cc(Cl)c(Cl)c(Cl)c1
Specifikationer
Kemiskt namn eller material 1,2,3-Trichloro-5-methylbenzene
Kvantitet 5 mg
Anteckningar om renhetsgrad GC
InChI formel InChI=1S/C7H5Cl3/c1-4-2-5(8)7(10)6(9)3-4/h2-3H,1H3
Formel vikt 193.9457
Procent renhet >95%