missing translation for 'onlineSavingsMsg'
Läs mer

1,2,3-Trichlorobenzene, 99%

Produktkod. p-7026247
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
500 g
Förpackningsstorlek:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet Förpackningsstorlek
11499677 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 11499677 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer B22154.36

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

CAS: 87-61-6 | C6H3Cl3 | 181.44 g/mol

Detoxification by catalytic hydrotreatment of 1,2,3-Trichlorobenzene is used for the disposal of hazardous organic waste liquids. As constituent of trichlorobenzene mixt used for termite control. As transformer fluid, dye carrier & solvent. Solvent for high melting products, Coolant in electrical installations and glass tempering. In polyester dyeing, lubricants, Heat transfer medium. As chemical intermediate for 2,3-dichlorophenol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Detoxification by catalytic hydrotreatment of 1,2,3-Trichlorobenzene is used for the disposal of hazardous organic waste liquids. As constituent of trichlorobenzene mixt used for termite control. As transformer fluid, dye carrier & solvent. Solvent for high melting products, Coolant in electrical installations and glass tempering. In polyester dyeing, lubricants, Heat transfer medium. As chemical intermediate for 2,3-dichlorophenol.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 87-61-6
Molekylformel C6H3Cl3
Molekylvikt (g/mol) 181.44
MDL-nummer MFCD00000537
InChI-nyckel RELMFMZEBKVZJC-UHFFFAOYSA-N
Synonym trichlorobenzene, vic-trichlorobenzene, 1,2,6-trichlorobenzene, benzene, 1,2,3-trichloro, benzene, trichloro, unii-nur9777ik4, ccris 5944, dsstox_cid_6193, dsstox_rid_78052, dsstox_gsid_26193
PubChem CID 6895
ChEBI CHEBI:35289
IUPAC-namn 1,2,3-trichlorobenzene
LEDER ClC1=CC=CC(Cl)=C1Cl

Specifikationer

Kemiskt namn eller material 1,2,3-Trichlorobenzene
Smältpunkt 53°C to 55°C
Kvantitet 500 g
UN-nummer UN3077
Beilstein 956882
Merck Index 14,9630
Kokpunkt 218°C to 219°C
Formel vikt 181.45
Procent renhet 99%

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.