Läs mer
1-[2-[(3S)-3-[3-[(1E)-2-(7-Chloro-2-quinolinyl)ethenyl]phenyl]-3-hydroxypropyl]phenyl]acetate, TRC
Produktkod. 30349033
Change view
Kvantitet:
25 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30349033 25 mg
30364608 50 mg
30353109 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30349033 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC37997525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 184764-13-4
Molekylformel C28 H24 Cl N O3
Molekylvikt (g/mol) 457.95
Synonym [S-(E)]-1-[2-[3-[3-[2-(7-Chloro-2-quinolinyl)ethenyl]phenyl]-3-hydroxypropyl]phenyl]ethanone
IUPAC-namn 2-[3-[(3S)-3-[3-[(E)-2-(7-chloroquinolin-2-yl)ethenyl]phenyl]-3-hydroxypropyl]phenyl]acetic acid
LEDER O[C@@H](CCc1cccc(CC(=O)O)c1)c2cccc(\C=C\c3ccc4ccc(Cl)cc4n3)c2
Specifikationer
Kemiskt namn eller material 1-[2-[(3S)-3-[3-[(1E)-2-(7-Chloro-2-quinolinyl)ethenyl]phenyl]-3-hydroxypropyl]phenyl]acetate
Kvantitet 25 mg
InChI formel InChI=1S/C28H24ClNO3/c29-24-11-9-22-10-13-25(30-26(22)18-24)12-7-20-4-2-6-23(16-20)27(31)14-8-19-3-1-5-21(15-19)17-28(32)33/h1-7,9-13,15-16,18,27,31H,8,14,17H2,(H,32,33)/b12-7+/t27-/m0/s1
Formel vikt 457.1445