missing translation for 'onlineSavingsMsg'
Learn More

1,2,4,5-Tetrafluorobenzene, 99%, Thermo Scientific Chemicals

Catalog Number p-7023849
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
327-54-8
C6H2F4
150.076
MFCD00000307
SDXUIOOHCIQXRP-UHFFFAOYSA-N
benzene, 1,2,4,5-tetrafluoro, 2,3,5,6-tetrafluorobenzene, pubchem1063, acmc-209hvc, 1,4,5-tetrafluorobenzene, 2,5,6-tetrafluorobenzene, 2,3,5,6-tetrafluorophenyl, ksc496i2b, benzene,2,4,5-tetrafluoro, 1,2,4,5-tetrafluoro-benzene
9474
1,2,4,5-tetrafluorobenzene
C1=C(C(=CC(=C1F)F)F)F
This item is not returnable. View return policy
327-54-8
C6H2F4
150.076
MFCD00000307
SDXUIOOHCIQXRP-UHFFFAOYSA-N
benzene, 1,2,4,5-tetrafluoro, 2,3,5,6-tetrafluorobenzene, pubchem1063, acmc-209hvc, 1,4,5-tetrafluorobenzene, 2,5,6-tetrafluorobenzene, 2,3,5,6-tetrafluorophenyl, ksc496i2b, benzene,2,4,5-tetrafluoro, 1,2,4,5-tetrafluoro-benzene
9474
1,2,4,5-tetrafluorobenzene
C1=C(C(=CC(=C1F)F)F)F

It is employed as intermediate in organic synthesis. It is also used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as intermediate in organic synthesis. It is also used as pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep away from ignition sources.
TRUSTED_SUSTAINABILITY
Melting Point 3°C to 5°C
Density 1.43
Boiling Point 90°C
Flash Point 16°C (60°F)
Refractive Index 1.407
Quantity 5 g
UN Number UN2387
Beilstein 1909052
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 150.07
Percent Purity 99%
Chemical Name or Material 1,2,4,5-Tetrafluorobenzene
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'