Läs mer
1-(2,4,6-Trihydroxyphenyl)-2-(4-methoxyphenyl)ethanone, TRC
Produktkod. 30452584
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30452584 50 mg
30454632 100 mg
30454315 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30452584 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT89580050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 15485-66-2
Molekylformel C15H14O5
Molekylvikt (g/mol) 274.26
IUPAC-namn 2-(4-methoxyphenyl)-1-(2,4,6-trihydroxyphenyl)ethanone
LEDER COc1ccc(CC(=O)c2c(O)cc(O)cc2O)cc1
Specifikationer
Kemiskt namn eller material 1-(2,4,6-Trihydroxyphenyl)-2-(4-methoxyphenyl)ethanone
Kvantitet 50 mg
InChI formel InChI=1S/C15H14O5/c1-20-11-4-2-9(3-5-11)6-12(17)15-13(18)7-10(16)8-14(15)19/h2-5,7-8,16,18-19H,6H2,1H3
Formel vikt 274.0841