missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
1-(2,4-Difluorophenyl)-2-(1H-1,2,4-triazol-1-yl)ethanol, Mikromol™
Description
1-(2,4-Difluorophenyl)-2-(1H-1,2,4-triazol-1-yl)ethanol
Specifications
Specifications
| Kemiskt namn eller material | 1-(2,4-Difluorophenyl)-2-(1H-1,2,4-triazol-1-yl)ethanol |
| CAS | 206050-23-9 |
| Analyt- eller komponentnamn | 1-(2,4-Difluorophenyl)-2-(1H-1,2,4-triazol-1-yl)ethanol |
| Frakt skick | Room Temperature |
| Molekylformel | C10 H9 F2 N3 O |
| Certifikat och efterlevnad | ISO 17025 |
| Synonym | 1-(2,4-Difluorophenyl)-2-(1H-1,2,4-triazol-1-yl)ethanol |
| InChI formel | InChI=1S/C10H9F2N3O/c11-7-1-2-8(9(12)3-7)10(16)4-15-6-13-5-14-15/h1-3,5-6,10,16H,4H2 |
| LEDER | OC(Cn1cncn1)c2ccc(F)cc2F |
| IUPAC-namn | 1-(2,4-difluorophenyl)-2-(1,2,4-triazol-1-yl)ethanol |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?