Läs mer
1-(2,4-Dihydroxyphenyl)-2-(4-hydroxyphenyl)propan-1-one, TRC
Produktkod. 30133215
Change view
Kvantitet:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133215 250 mg
30133238 500 mg
17919094 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30133215 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD454245250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 21255-69-6
Molekylformel C15H14O4
Molekylvikt (g/mol) 258.27
Synonym O-Desmethylangolensin
LEDER O=C(C(C)C1=CC=C(O)C=C1)C2=C(O)C=C(O)C=C2
Specifikationer
Kemiskt namn eller material 1-(2,4-Dihydroxyphenyl)-2-(4-hydroxyphenyl)propan-1-one
Anteckningar om renhetsgrad HPLC
Kvantitet 250 mg
InChI formel InChI=1S/C15H14O4/c1-9(10-2-4-11(16)5-3-10)15(19)13-7-6-12(17)8-14(13)18/h2-9,16-18H,1H3
Formel vikt 258.0892
Procent renhet >95%