Läs mer
1-[2-(4-Hydroxy-2,2,6,6-tetramethyl-1-piperidinyl)ethyl] Ester Butanedioic Acid, TRC
Produktkod. 30012752
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30012752 100 mg
30012127 10 mg
30011865 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30012752 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH371430100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1872428-97-1
Molekylformel C15 H27 N O5
Molekylvikt (g/mol) 301.379
IUPAC-namn 4-[2-(4-hydroxy-2,2,6,6-tetramethylpiperidin-1-yl)ethoxy]-4-oxobutanoic acid
LEDER CC1(C)CC(O)CC(C)(C)N1CCOC(=O)CCC(=O)O
Specifikationer
Kemiskt namn eller material 1-[2-(4-Hydroxy-2,2,6,6-tetramethyl-1-piperidinyl)ethyl] Ester Butanedioic Acid
Kvantitet 100 mg
InChI formel InChI=1S/C15H27NO5/c1-14(2)9-11(17)10-15(3,4)16(14)7-8-21-13(20)6-5-12(18)19/h11,17H,5-10H2,1-4H3,(H,18,19)
Formel vikt 301.189