Läs mer
1,2,4-Triacetoxybenzene, TRC
Produktkod. 30449689
Change view
Kvantitet:
25 g
Produktkod. Kvantitet
30449689 25 g
1 options
Produktkod. 30449689 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT72085525G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1,2,4-Triacetoxybenzene
CAS 613-03-6
Molekylformel C12H12O6
Kvantitet 25 g
Synonym 1,2,4-Tris(acetoxy)benzene, 2-Hydroxyhydroquinone triacetateHydroxyhydroquinone triacetate, Pyrogallol A, NSC 2149
InChI formel InChI=1S/C12H12O6/c1-7(13)16-10-4-5-11(17-8(2)14)12(6-10)18-9(3)15/h4-6H,1-3H3
LEDER CC(=O)Oc1ccc(OC(=O)C)c(OC(=O)C)c1
IUPAC-namn (3,4-diacetyloxyphenyl) acetate
Molekylvikt (g/mol) 252.22
Formel vikt 252.0634