Läs mer
[1,2,4]Triazolo[1,5-a]pyridin-5-ol, TRC
Produktkod. 30427649
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30427649 10 mg
30415575 50 mg
30409450 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30427649 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT13945010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1824144-99-1
Molekylformel C6H5N3O
Molekylvikt (g/mol) 135.12
IUPAC-namn [1,2,4]triazolo[1,5-a]pyridin-5-ol
LEDER Oc1cccc2ncnn12
Specifikationer
Kemiskt namn eller material [1,2,4]Triazolo[1,5-a]pyridin-5-ol
Kvantitet 10 mg
InChI formel InChI=1S/C6H5N3O/c10-6-3-1-2-5-7-4-8-9(5)6/h1-4,10H
Formel vikt 135.0433