Läs mer
1,2,4-Trinonyl Ester 1,2,4-Benzenetricarboxylic Acid, TRC
Produktkod. 30450660
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30450660 100 mg
1 options
Produktkod. 30450660 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT886290100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Trinonyl benzene-1,2,4-tricarboxylate
CAS 35415-27-1
Molekylformel C36 H60 O6
Kvantitet 100 mg
Synonym 1,2,4-Benzenetricarboxylic Acid Trinonyl Ester, Trimellitic Acid Trinonyl Ester, Tri-n-nonyl Trimellitate, Trinonyl Trimellitate
InChI formel InChI=1S/C36H60O6/c1-4-7-10-13-16-19-22-27-40-34(37)31-25-26-32(35(38)41-28-23-20-17-14-11-8-5-2)33(30-31)36(39)42-29-24-21-18-15-12-9-6-3/h25-26,30H,4-24,27-29H2,1-3H3
LEDER CCCCCCCCCOC(=O)c1ccc(C(=O)OCCCCCCCCC)c(c1)C(=O)OCCCCCCCCC
IUPAC-namn trinonyl benzene-1,2,4-tricarboxylate
Molekylvikt (g/mol) 588.858
Formel vikt 588.439