Läs mer
1,2,4-Trioctyl Ester 1,2,4-Benzenetricarboxylic Acid, TRC
Produktkod. 30134775
Change view
Kvantitet:
250 mg
2.5 g
Förpackningsstorlek:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30134775 2.5 g 2.5g
30004414 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30134775 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT8863002.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 89-04-3
Molekylformel C33H54O6
Molekylvikt (g/mol) 546.78
Synonym Trimellitic Acid Trioctyl Ester, ADK Cizer C 8, Adekacizer C 8, Crodamol TOTM, Diplast TM 8, Monocizer W 750, PX 338, Tri-n-octyl Trimellitate, Trimellitate N 08, Trimex N 08, Trioctyl 1,2,4-Benzenetricarboxylate, Trioctyl Trimellitate, Tris(n-octyl)Trimellitate, Tris(octyl) Trimellitate, Vinycizer W 700, W 700, W 750, W 755
IUPAC-namn trioctyl benzene-1,2,4-tricarboxylate
LEDER CCCCCCCCOC(=O)c1ccc(C(=O)OCCCCCCCC)c(c1)C(=O)OCCCCCCCC
Specifikationer
Kemiskt namn eller material 1,2,4-Trioctyl Ester 1,2,4-Benzenetricarboxylic Acid
Kvantitet 2.5 g
InChI formel InChI=1S/C33H54O6/c1-4-7-10-13-16-19-24-37-31(34)28-22-23-29(32(35)38-25-20-17-14-11-8-5-2)30(27-28)33(36)39-26-21-18-15-12-9-6-3/h22-23,27H,4-21,24-26H2,1-3H3
Formel vikt 546.392