Läs mer
1,2:5,6-Di-O-cyclohexylidene-myo-inositol, TRC
Produktkod. 30370955
Change view
Kvantitet:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30370955 5 g
30370102 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30370955 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD4390105G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 34711-26-7
Molekylformel C18H28O6
Molekylvikt (g/mol) 340.41
Synonym (±)-2,3:4,5-Di-O-cyclohexylidene-D-myo-inositol, 1,2:5,6-Di-O-cyclohexylidene-DL-myo-inositol, DL-Myo-1,2:5,6-di-O-cyclohexylideneinositol
LEDER O[C@@H]1[C@@H](O)[C@@H]2OC3(CCCCC3)O[C@H]2[C@@H]4OC5(CCCCC5)O[C@H]14
Specifikationer
Kemiskt namn eller material 1,2:5,6-Di-O-cyclohexylidene-myo-inositol
Kvantitet 5 g
InChI formel InChI=1S/C18H28O6/c19-11-12(20)14-16(24-18(22-14)9-5-2-6-10-18)15-13(11)21-17(23-15)7-3-1-4-8-17/h11-16,19-20H,1-10H2/t11-,12-,13-,14+,15-,16-/m1/s1
Formel vikt 340.1886