Learn More
Alfa Aesar™ 1-(2,5-Dichlorophenyl)-3-[4-(trifluoromethoxy)phenyl]urea, 97%
Brand:  Alfa Aesar™ H64182.06
Additional Details : Weight : 0.00500kg
Quantity 5g
 View more versions of this product
Product Code. 15498746
  • 11791.55 SEK / 5g
Estimated Shipment date
from Supplier 02-02-2018
Add to basket
Specifications
Chemical Identifiers
1-(2,5-Dichlorophenyl)-3-[4-(trifluoromethoxy)phenyl]urea
365.13
MFCD04281091
Specifications
1-(2,5-Dichlorophenyl)-3-[4-(trifluoromethoxy)phenyl]urea
Solid
365.13
5g
97%
MFCD04281091