missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(2,5-Dimethylphenyl)-3-[4-(trifluoromethyl)phenyl]urea, 97%

Brand:  Alfa Aesar™ H61354.03

Additional Details : Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15486456

  • 6228.40 SEK / 1g


Estimated Shipment date
from Supplier 04-07-2018
Add to basket

Specifications

Chemical Identifiers

1-(2,5-Dimethylphenyl)-3-[4-(trifluoromethyl)phenyl]urea
308.3
MFCD05080089

Specifications

1-(2,5-Dimethylphenyl)-3-[4-(trifluoromethyl)phenyl]urea
Solid
308.3
1g
97%
MFCD05080089
Videos
SDS