Läs mer
1-(2,6-Dichlorophenyl)-1,3-dihydro-2H-indol-2-one, Mikromol™
Produktkod. 17944226
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17944226 100 mg
17991956 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17944226 Leverantör Mikromol Leverantörsnummer MM0006.01
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1-(2,6-Dichlorophenyl)-1,3-dihydro-2H-indol-2-one
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1-(2,6-Dichlorophenyl)-1,3-dihydro-2H-indol-2-one
CAS 15362-40-0
Analyt- eller komponentnamn 1-(2,6-Dichlorophenyl)-1,3-dihydro-2H-indol-2-one
Frakt skick Room Temperature
Molekylformel C14 H9 Cl2 N O
Certifikat och efterlevnad ISO 17025
Synonym 1-(2,6-Dichlorophenyl)-1,3-dihydro-2H-indol-2-one, Aceclofenac Imp. I (EP), Diclofenac Sodium Imp. A (EP), Diclofenac Related Compound A (USP), Diclofenac Related Compound A, Aceclofenac Impurity I, Diclofenac Sodium Impurity A, Diclofenac Impurity A
InChI formel InChI=1S/C14H9Cl2NO/c15-10-5-3-6-11(16)14(10)17-12-7-2-1-4-9(12)8-13(17)18/h1-7H,8H2
LEDER Clc1cccc(Cl)c1N2C(=O)Cc3ccccc23
IUPAC-namn 1-(2,6-dichlorophenyl)-3H-indol-2-one
Molekylvikt (g/mol) 278.13
Kvantitet 100 mg
Formel vikt 277.0061 g/mol