missing translation for 'onlineSavingsMsg'
Learn More

1,2,6-Hexanetriol, 97+%, extra pure, Thermo Scientific Chemicals

Catalog Number p-142328
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
106-69-4
C6H14O3
134.18
MFCD00002976
ZWVMLYRJXORSEP-UHFFFAOYNA-N
1,2,6-hexanetriol, 1,2,6-trihydroxyhexane, hexanetriol-1,2,6, 1,6-hexanetriol, hexanetriol-1,6, hexane-1,6-triol, 1,6-trihydroxyhexane, acmc-1bsub, dsstox_cid_21224
7823
hexane-1,2,6-triol
OCCCCC(O)CO
This item is not returnable. View return policy
106-69-4
C6H14O3
134.18
MFCD00002976
ZWVMLYRJXORSEP-UHFFFAOYNA-N
1,2,6-hexanetriol, 1,2,6-trihydroxyhexane, hexanetriol-1,2,6, 1,6-hexanetriol, hexanetriol-1,6, hexane-1,6-triol, 1,6-trihydroxyhexane, acmc-1bsub, dsstox_cid_21224
7823
hexane-1,2,6-triol
OCCCCC(O)CO

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.0
CAS Max % 100.0
Melting Point 25.0°C to 32.0°C
Color Yellow
Density 1.1090g/mL
pH 4.0 to 7.0 (30% soln. at 20°C)
Boiling Point 178.0°C (6.7 mmHg)
Flash Point 198°C
Infrared Spectrum Authentic
Assay Percent Range 97% min. (GC)
Packaging Glass bottle
Linear Formula HO(CH2)4CH(OH)CH2OH
Refractive Index 1.4750 to 1.4770
Quantity 100 g
Beilstein 01,IV,2784
Specific Gravity 1.109
Solubility Information Solubility in water: miscible with water. Other solubilities: miscible with acetone and alcohol, immiscible with benzene, toluene, heptane and, diethyl ether, practically insoluble in ether, low solubility in non-polar solvents
Formula Weight 134.18
Percent Purity 97+%
Grade Extra Pure
Physical Form Viscous Liquid
Chemical Name or Material 1, 2, 6-Hexanetriol, 97%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'