Learn More
1,2,7,8-Diepoxyoctane, 97%, Thermo Scientific Chemicals
Catalog Number p-7044496
737.67 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
10 g
50 g
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
2426-07-5
C8H14O2
142.20
MFCD00005155
LFKLPJRVSHJZPL-UHFFFAOYNA-N
1,2,7,8-diepoxyoctane, 1,2:7,8-diepoxyoctane, 1,7-octadiene diepoxide, oxirane, 2,2'-1,4-butanediyl bis, 1,2-epoxy-7,8-epoxyoctane, 2,2'-1,4-butanediyl bisoxirane, 1,4-di oxiran-2-yl butane, octane, 1,2:7,8-diepoxy, 2-4-oxiran-2-yl butyl oxirane, ccris 963
17048
CHEBI:23705
2-[4-(oxiran-2-yl)butyl]oxirane
C1C(O1)CCCCC2CO2
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
2426-07-5
C8H14O2
142.20
MFCD00005155
LFKLPJRVSHJZPL-UHFFFAOYNA-N
1,2,7,8-diepoxyoctane, 1,2:7,8-diepoxyoctane, 1,7-octadiene diepoxide, oxirane, 2,2'-1,4-butanediyl bis, 1,2-epoxy-7,8-epoxyoctane, 2,2'-1,4-butanediyl bisoxirane, 1,4-di oxiran-2-yl butane, octane, 1,2:7,8-diepoxy, 2-4-oxiran-2-yl butyl oxirane, ccris 963
17048
CHEBI:23705
2-[4-(oxiran-2-yl)butyl]oxirane
C1C(O1)CCCCC2CO2
1,2,7,8-Diepoxyoctane is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,2,7,8-Diepoxyoctane is used as an organic chemical synthesis intermediate.
Solubility
Soluble in water (partly miscible), acetone and heptane.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Density 0.997
Boiling Point 238°C to 240°C
Flash Point 97°C (206°F)
Refractive Index 1.445
Quantity 10 g
UN Number UN2922
Beilstein 104873
Solubility Information Soluble in water (partly miscible),acetone and heptane.
Formula Weight 142.2
Percent Purity 97%
Chemical Name or Material 1,2,7,8-Diepoxyoctane