missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(2-Amino-5-chlorophenyl)-2,2,2-trifluoroethanone, Mikromol™
Description
1-(2-Amino-5-chlorophenyl)-2,2,2-trifluoroethanone
Specifications
Specifications
| Kemiskt namn eller material | 4-Chloro-2-trifluoroacetylaniline |
| CAS | 154598-53-5 |
| Analyt- eller komponentnamn | 4-Chloro-2-trifluoroacetylaniline |
| Frakt skick | Room Temperature |
| Molekylformel | C8 H5 Cl F3 N O |
| Certifikat och efterlevnad | ISO 9001 |
| Synonym | 1-(2-Amino-5-chlorophenyl)-2,2,2-trifluoroethanone |
| InChI formel | InChI=1S/C8H5ClF3NO/c9-4-1-2-6(13)5(3-4)7(14)8(10,11)12/h1-3H,13H2 |
| LEDER | Nc1ccc(Cl)cc1C(=O)C(F)(F)F |
| IUPAC-namn | 1-(2-amino-5-chlorophenyl)-2,2,2-trifluoroethanone |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?