Läs mer
1-[[2'-(Aminocarbonyl)[1,1'-biphenyl]-4-yl]methyl]-2-ethoxy-1H-benzimidazole-7-carboxylic Acid Methyl Ester, TRC
Produktkod. 30319948
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30319948 250 mg
1 options
Produktkod. 30319948 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA147405250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Methyl 1-((2'-Carbamoyl-[1,1'-biphenyl]-4-yl)methyl)-2-ethoxy-1H-benzo[d]imidazole-7-carboxylate
CAS 147404-76-0
Molekylformel C25 H23 N3 O4
Kvantitet 250 mg
Synonym 1-[[2'-(Aminocarbonyl)[1,1'-biphenyl]-4-yl]methyl]-2-ethoxy-1H-benzimidazole-7-carboxylic Acid Methyl Ester, Azilsartan impurity A
InChI formel InChI=1S/C25H23N3O4/c1-3-32-25-27-21-10-6-9-20(24(30)31-2)22(21)28(25)15-16-11-13-17(14-12-16)18-7-4-5-8-19(18)23(26)29/h4-14H,3,15H2,1-2H3,(H2,26,29)
LEDER CCOc1nc2cccc(C(=O)OC)c2n1Cc3ccc(cc3)c4ccccc4C(=O)N
IUPAC-namn methyl 3-[[4-(2-carbamoylphenyl)phenyl]methyl]-2-ethoxy-benzimidazole-4-carboxylate
Molekylvikt (g/mol) 429.47
Formel vikt 429.1689