Läs mer
1,2-Benzenedicarboxylic Acid, mixed decyl and hexyl and octyl diesters (1:1:1 Mixture of B185420 and, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Phthalic acid, mixed decyl-hexyl-octyl diester |
| CAS | 68648-93-1 |
| Analyt- eller komponentnamn | Phthalic acid, mixed decyl-hexyl-octyl diester |
| Anteckningar om renhetsgrad | HPLC |
| Frakt skick | Room Temperature |
| Molekylformel | C12 H14 O4 |
| Synonym | Phthalic acid, mixed decyl-hexyl-octyl diester, 1,2-Benzenedicarboxylic acid, mixed decyl and hexyl and octyl diesters, 1,2-Benzenedicarboxylic acid, di-C6-10-alkyl esters |
| Lösningstyp | Neat |
| LEDER | O=C(OCCCCCCCC)C1=CC=CC=C1C(OCCCCCC)=O.O=C(OCCCCCCCCCC)C2=CC=CC=C2C(OCCCCCC)=O.O=C(OCCCCCCCCCC)C3=CC=CC=C3C(OCCCCCCCC)=O |
| Industry Type | Forensics |
| Visa mer |