Läs mer
1,2-Benzenedicarboxylic Acid, Mixed Decyl, Hexyl and Octyl Diesters(1:1:1 Mixture of B185420 and D228455 and D228460), TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Phthalic acid, mixed decyl-hexyl-octyl diester |
| CAS | 68648-93-1 |
| Anteckningar om renhetsgrad | HPLC |
| Molekylformel | C12 H14 O4 |
| Kvantitet | 250 mg |
| Synonym | Phthalic acid, mixed decyl-hexyl-octyl diester, 1,2-Benzenedicarboxylic acid, mixed decyl and hexyl and octyl diesters, 1,2-Benzenedicarboxylic acid, di-C6-10-alkyl esters |
| LEDER | O=C(OCCCCCCCC)C1=CC=CC=C1C(OCCCCCC)=O.O=C(OCCCCCCCCCC)C2=CC=CC=C2C(OCCCCCC)=O.O=C(OCCCCCCCCCC)C3=CC=CC=C3C(OCCCCCCCC)=O |
| Molekylvikt (g/mol) | 222.23 |
| Formel vikt | 222.0892 |
| Procent renhet | >95 |