missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,2-Benzenedimethanol, 97%, Thermo Scientific Chemicals
386.58 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 g
5 g
25 g
Description
1,2-Benzenedimethanol is a raw material used in chemical synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,2-Benzenedimethanol is a raw material used in chemical synthesis.
Solubility
Soluble in water, ether, ethanol, benzene.
Notes
Store in cool, dry conditions in a sealed container. Incompatible with oxidizing agents.
1,2-Benzenedimethanol is a raw material used in chemical synthesis.
Solubility
Soluble in water, ether, ethanol, benzene.
Notes
Store in cool, dry conditions in a sealed container. Incompatible with oxidizing agents.
Specifications
Melting Point | 59°C to 64°C |
Quantity | 1 g |
Beilstein | 508775 |
Solubility Information | Soluble in water,ether,ethanol,benzene. |
Formula Weight | 138.17 |
Percent Purity | 97% |
Chemical Name or Material | 1,2-Benzenedimethanol |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'