missing translation for 'onlineSavingsMsg'
Läs mer

1,2-Benzisoxazole-3-methanesulfonate Sodium Salt, TRC

Produktkod. 30010756
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
1 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30010756 50 g
30138265 1 g
30132874 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30010756 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB20195050G

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73101-64-1
Kemiskt namn eller material 1,2-Benzisoxazole-3-methanesulfonate, Sodium Salt
Formel vikt 234.9915
InChI formel InChI=1S/C8H7NO4S.Na/c10-14(11,12)5-7-6-3-1-2-4-8(6)13-9-7;/h1-4H,5H2,(H,10,11,12);/q;+1/p-1
IUPAC-namn sodium;1,2-benzoxazol-3-ylmethanesulfonate
Molekylformel C8 H6 N O4 S . Na
Molekylvikt (g/mol) 235.19
LEDER [Na+].[O-]S(=O)(=O)Cc1noc2ccccc12
Synonym 1,2-Benzisoxazole-3-methanesulfonic acid, sodium salt (1:1), 1,2-Benzisoxazole-3-methanesulfonic acid sodium salt, 1,2-Benzisoxazole-3-methanesulfonic acid, sodium salt (9CI), Sodium 1,2-benzisoxazole-3-methanesulfonate

Specifications

Kvantitet 50 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.