Läs mer
1,2-Bis(2,4,6-tribromophenoxy)ethane, TRC
Produktkod. 30135327
Change view
Kvantitet:
500 mg
1 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135327 1 g
30132493 10 g
17902895 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30135327 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB5854251G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 37853-59-1
Molekylformel C14 H8 Br6 O2
Molekylvikt (g/mol) 687.64
Synonym 1,1'-[1,2-Ethanediylbis(oxy)]bis[2,4,6-tribromobenzene], 1,2-Bis(2,4,6-tribromophenoxy)ethane, BTBPE, FF 680, FI 680, FM 680, FireMaster 680, FireMaster FF 680
IUPAC-namn 1,3,5-tribromo-2-[2-(2,4,6-tribromophenoxy)ethoxy]benzene
LEDER Brc1cc(Br)c(OCCOc2c(Br)cc(Br)cc2Br)c(Br)c1
Specifikationer
Kemiskt namn eller material 1,2-Bis(2,4,6-tribromophenoxy)ethane
Kvantitet 1 g
InChI formel InChI=1S/C14H8Br6O2/c15-7-3-9(17)13(10(18)4-7)21-1-2-22-14-11(19)5-8(16)6-12(14)20/h3-6H,1-2H2
Formel vikt 681.5625