Läs mer
1,2-Bis(7-methyloctyl)cyclohexyl-1,2-dicarboxylate, TRC
Produktkod. 30180010
Change view
Kvantitet:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30180010 10 g
30180092 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30180010 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB49610010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 318292-43-2
Molekylformel C26 H48 O4
Molekylvikt (g/mol) 424.66
Synonym 1,2-Cyclohexanedicarboxylic acid, bis(7-methyloctyl) ester (9CI), Bis(7-methyloctyl) cyclohexane-1,2-dicarboxylate, Bis(7-methyloctyl) tetrahydrophthalate
IUPAC-namn bis(7-methyloctyl) cyclohexane-1,2-dicarboxylate
LEDER CC(C)CCCCCCOC(=O)C1CCCCC1C(=O)OCCCCCCC(C)C
Specifikationer
Kemiskt namn eller material 1,2-Cyclohexanedicarboxylic acid, bis(7-methyloctyl) ester
Kvantitet 10 g
InChI formel InChI=1S/C26H48O4/c1-21(2)15-9-5-7-13-19-29-25(27)23-17-11-12-18-24(23)26(28)30-20-14-8-6-10-16-22(3)4/h21-24H,5-20H2,1-4H3
Formel vikt 424.3553