missing translation for 'onlineSavingsMsg'
Learn More

1,2-Bis(chlorodimethylsilyl)ethane, tech. 90%, Thermo Scientific Chemicals

Catalog Number p-7043758
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
13528-93-3
C6H16Cl2Si2
215.26
MFCD00000505
VGQOKOYKFDUPPJ-UHFFFAOYSA-N
1,2-bis chlorodimethylsilyl ethane, ethylenebis chlorodimethylsilane, silane, 1,2-ethanediylbis chlorodimethyl, unii-b4ocj0e6j1, 1,1,4,4-tetramethyl-1,4-dichlorodisilethylene, chloro 2-chlorodimethylsilyl ethyl dimethylsilane, b4ocj0e6j1, 2,5-dichloro-2,5-dimethyl-2,5-disilahexane, 1,1,4,4-tetramethyl-1,4-dichloro-disylethylene
83552
C[Si](C)(Cl)CC[Si](C)(C)Cl
This item is not returnable. View return policy
13528-93-3
C6H16Cl2Si2
215.26
MFCD00000505
VGQOKOYKFDUPPJ-UHFFFAOYSA-N
1,2-bis chlorodimethylsilyl ethane, ethylenebis chlorodimethylsilane, silane, 1,2-ethanediylbis chlorodimethyl, unii-b4ocj0e6j1, 1,1,4,4-tetramethyl-1,4-dichlorodisilethylene, chloro 2-chlorodimethylsilyl ethyl dimethylsilane, b4ocj0e6j1, 2,5-dichloro-2,5-dimethyl-2,5-disilahexane, 1,1,4,4-tetramethyl-1,4-dichloro-disylethylene
83552
C[Si](C)(Cl)CC[Si](C)(C)Cl

1,2-Bis(chlorodimethylsilyl)ethane is used as Protecting Reagent for Primary Amines. It is also used in Medicine field, electronics industry and polymer material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Bis(chlorodimethylsilyl)ethane is used as Protecting Reagent for Primary Amines. It is also used in Medicine field, electronics industry and polymer material.

Solubility
Reacts with water.

Notes
Moisture Sensitive. Incompatible with Strong oxidizing agents, Strong bases and water. Store under dry inert gas.
TRUSTED_SUSTAINABILITY
Boiling Point 198°C to 200°C
Melting Point 34°C to 40°C
Quantity 25 g
UN Number UN3261
Beilstein 1920141
Flash Point 65°C (149°F)
Solubility Information Reacts with water.
IUPAC Name chloro-[2-[chloro(dimethyl)silyl]ethyl]-dimethylsilane
Formula Weight 215.27
Percent Purity 90%
Grade Technical
Sensitivity Moisture sensitive
Chemical Name or Material 1,2-Bis(chlorodimethylsilyl)ethane
Show More Show Less
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'