Läs mer
1,2-Bis(m-tolyloxy)ethane, TRC
Produktkod. 30483564
Change view
Kvantitet:
1 g
2.5 g
5 g
Förpackningsstorlek:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30483564 2.5 g 2.5g
30482911 1 g -
30471865 5 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30483564 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3333482.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 54914-85-1
Molekylformel C16H18O2
Molekylvikt (g/mol) 242.31
Synonym 1,1'-[1,2-Ethanediylbis(oxy)]bis[3-methylbenzene], 1,1'-[ethane-1,2-diylbis(oxy)]bis(3-methylbenzene), 1,2-Bis(3-methylphenoxy)ethane, 1,2-Bis(3'-methylphenoxy)ethane, 1,2-Di(3-methylphenoxy)ethane, 1,2-Di(m-tolyloxy)ethane, 1-Methyl-3-[2-(3-methylphenoxy)ethoxy]benzene, Ethylene glycol bis(m-tolyl ether), KS 232, NSC 117525
IUPAC-namn 1-methyl-3-[2-(3-methylphenoxy)ethoxy]benzene
LEDER Cc1cccc(OCCOc2cccc(C)c2)c1
Specifikationer
Kemiskt namn eller material 1,2-Bis(m-tolyloxy)ethane
Kvantitet 2.5 g
InChI formel InChI=1S/C16H18O2/c1-13-5-3-7-15(11-13)17-9-10-18-16-8-4-6-14(2)12-16/h3-8,11-12H,9-10H2,1-2H3
Formel vikt 242.1307