Läs mer
1-[(2'-Carbamoylbiphenyl-4-yl) methyl]-2-ethoxybenzimidazole-7-carboxylic Acid, TRC
Produktkod. 30362099
Change view
Kvantitet:
10 mg
50 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30362099 50 mg
30360593 500 mg
30357526 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30362099 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD22920650MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1696392-11-6
Molekylformel C24 H21 N3 O4
Molekylvikt (g/mol) 415.44
Synonym 1-[[2'-(Aminocarbonyl)[1,1'-biphenyl]-4-yl]methyl]-2-ethoxy-1H-benzimidazole-7-carboxylic Acid
IUPAC-namn 3-[[4-(2-carbamoylphenyl)phenyl]methyl]-2-ethoxy-benzimidazole-4-carboxylic acid
LEDER CCOc1nc2cccc(C(=O)O)c2n1Cc3ccc(cc3)c4ccccc4C(=O)N
Specifikationer
Kemiskt namn eller material 1-[(2'-Carbamoylbiphenyl-4-yl)methyl]-2-ethoxybenzimidazole-7-carboxylic acid
Kvantitet 50 mg
InChI formel InChI=1S/C24H21N3O4/c1-2-31-24-26-20-9-5-8-19(23(29)30)21(20)27(24)14-15-10-12-16(13-11-15)17-6-3-4-7-18(17)22(25)28/h3-13H,2,14H2,1H3,(H2,25,28)(H,29,30)
Formel vikt 415.1532